missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine sulfate, 99+%, Thermo Scientific Chemicals

Catalog Number p-143740
27.46 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10034-93-2
H4N2·H2SO4
130.12
MFCD00044873
ZGCHATBSUIJLRL-UHFFFAOYSA-N
hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate
24842
hydrazine;sulfuric acid
NN.OS(=O)(=O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
10034-93-2
H4N2·H2SO4
130.12
MFCD00044873
ZGCHATBSUIJLRL-UHFFFAOYSA-N
hydrazine sulfate, hydrazine monosulfate, hydrazine, sulfate, hydrazine sulphate, hydrazinium sulfate, hydrazonium sulfate, hydrazine sulfate 1:1, segidrin, sehydrin, diamine sulfate
24842
hydrazine;sulfuric acid
NN.OS(=O)(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Reduction reagent
TRUSTED_SUSTAINABILITY
Melting Point 254.0°C
Color White
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula H2NNH2·H2SO4
Quantity 100 g
Merck Index 15, 4810
Solubility Information Solubility in water: 30g/L (20°C). Other solubilities: insoluble in alcohol
Formula Weight 130.12
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Hydrazine sulfate, p.a.
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
  • P363-Wash contaminated clothing before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'