Learn More
Hydrochloric Acid, 0.5M, Reagecon™
BIOPHARMA
Product Code. 11526773 Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Packaging:
1L
Unit Size:
1 L
Product Code. 11526773
Brand: Reagecon H20501
Specifications
Concentration 0.5M
Packaging 1L
Quantity 1 L
compliance-icons
Product Identifier
  • HYDROCHLORIC ACID 0.5N (0.5M)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : hydrochloric acid