missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen hexafluoroantimonate(V), ≈65% aq. soln., Thermo Scientific Chemicals

Catalog Number p-7050031
94.67 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 g
50 g
100 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
16950-06-4
MFCD00011327
Hexafluoroantimonic acid; Fluoroantimonic acid
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
16950-06-4
MFCD00011327
Hexafluoroantimonic acid; Fluoroantimonic acid

Hydrogen hexafluoroantimonate(V) is an excellent medium for the preparation of alkylidene oxonium salts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Orange
Linear Formula HF·SbF5
Quantity 25 g
UN Number UN3264
Solubility Information It is fully miscible with water,soluble in SO2ClF,SO2.
Formula Weight 236.75
Concentration or Composition (by Analyte or Components) ≈65% aq. soln.
Physical Form Liquid
Chemical Name or Material Hydrogen hexafluoroantimonate(V)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'