missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen perOxide Solution, 50 wt% in H2O, Honeywell Fluka™

Catalog Number p-7072213
Click to view available options
Quantity:
500 mL
4 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7722-84-1
H2O2
34.014
MFCD00011333
MHAJPDPJQMAIIY-UHFFFAOYSA-N
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
784
CHEBI:16240
hydrogen peroxide
OO
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
7722-84-1
H2O2
34.014
MFCD00011333
MHAJPDPJQMAIIY-UHFFFAOYSA-N
oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
784
CHEBI:16240
hydrogen peroxide
OO

50 wt. % in H2O, stabilized

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
UN Number UN2014
Beilstein 3587191
Density 1.197g/mL at 20°C
Formula Weight 34.01g/mol
Vapor Pressure 23.3 mmHg (30°C)
Quantity 500 mL
Chemical Name or Material Hydrogen peroxide solution
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrogen peroxide solution
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 4
  • Oxidising liquid Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
  • H413-May cause long lasting harmful effects to aquatic life.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : hydrogen peroxide solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'