missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen peroxide solution, Honeywell™
BIOPHARMA

Catalog Number p-7145542
Click to view available options
Quantity:
1 L
5 L
4 x 5 L
6 x 1 L
This item is not returnable. View return policy
7732-18-5, 7722-84-1
H2O
18.02
XLYOFNOQVPJJNP-UHFFFAOYSA-N
784
CHEBI:16240
water
O
This item is not returnable. View return policy
7732-18-5, 7722-84-1
H2O
18.02
XLYOFNOQVPJJNP-UHFFFAOYSA-N
784
CHEBI:16240
water
O

Meets analytical specification of Ph. Nord., 34.5-36.5%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY
CAS Min % ≥50.00%, ≥30.00%
CAS Max % <70.00%, <50.00%
Packaging Plastic Bottle
Quantity 1 L
Physical Form Liquid
Grade Reagent pharmacopeia chemistry reagents
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'