Learn More
Honeywell™ HYDROGRAIN Gloves
Honeywell™ HYDROGRAIN Gloves designed for handling indoor and outdoor operations in a damp or greasy environment.
Brand: Honeywell™ 2049260-08
Additional Details : Weight : 0.01000kg
Description
- 5 finger leather glove enriched by a special tanning process to make the glove more water repellent
- Gunn cut wrist type
- Winged thumb makes the glove more anatomical for greater ease of use
- Fully water-repellent grain cowhide leather
- Elasticated wrist offers added support
- Outstanding comfort and flexibility
Specifications
0.8 to 1.1 mm | |
8 | |
25 cm |
Cowhide Leather | |
Yellow | |
EN 420-2003 EEC Basic standards, EN 388-2016 Mechanical hazards |