missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Honeywell™ Hydrograin Grain Cowhide Leather Gloves
Click to view available options
Size:
8
9
11
Description
- Fully water-repellent grain cowhide leather
- For handling operations in a damp or greasy environment
- Ideal for indoor and outdoor applications
- Elasticated wrist
- Gunn cut
- Wing thumb
Specifications
Certifications/Compliance | EN 420-2003 EEC Basic standards, EN 388-2016 Mechanical hazards |
Certifications/Compliance (Europe) | EN 420-2003 EEC Basic Standards, EN 388-2016 Mechanical Hazards |
Coating Material | Leather |
Color | Yellow |
Cut Resistance Rating | 1 |
Hand | Specific |
Material | Grain Cowhide Leather |
Product Type | Gloves |
Size | 8 |
Style | Gunn |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'