Learn More
Fisherbrand™ Hydrometer
Click to view available options
Specific Gravity Range:
0 to 100% NaCl Saturation
0 to 26.5% NaCl Saturation
1000 to 2000 g/L
1400 to 1620 g/L
1600 to 1820 g/L
1720 to 1790 g/L
1800 to 2000 g/L
2000 to 2500 g/L
2500 to 3000 g/L
650 to 1000 g/L
795 to 910 g/L
895 to 1000 g/L
Unit Size:
Each
Specifications
Specifications
| Quantity | 1Each |
| Type | Hydrometer |
| Length (Metric) | 150 mm |
| For Use With (Application) | For general use (laboratory, education, industry) |
| Specific Gravity Range | 650 to 1000 g/L |