Learn More
Merck Hydrosol Stainless Steel Filter Holder and Accessories
Accessories for filter holders for sample preparation.
Brand:  Merck XX2004702
1036.68 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10450103
  • 1115.00€ / Each
Estimated Shipment: 29-07-2024
to see stock.
Add to basket
Description
Description
Accessories for filter holders for sample preparation.
The Hydrosol filter holder replacement PartsXX2004704 Funnel and Locking Ring Assembly, stainless steelXX2004702 Base with Screen, gasket and supportXX20047RK Replacement Parts Kit for Hydrosol Filter Holder
Specifications
Specifications
Not Autoclavable
47 mm hydrosol vacuum filter
Non-sterile
Stainless steel base and filter screen, silicone stopper, PTFE gasket
Stainless Steel
47 mm