Learn More
Ansell HyFlex™ 11-130 Seamless Knit Polyamide Antistatic Gloves with Coated Palm
Product Code. 16039891 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
Product Code. 16039891
Brand: Ansell 11130090
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Gloves are knitted using a seamless, non-stop process with high-tenacity polyamid continuous thread. Antistatic properties protect users from electrostatic discharge.
  • Gloves are knitted using a seamless, non-stop process
  • Textile liner made of high-tenacity polyamid continuous thread that includes copper
  • Seamless, fine stitches enhance user comfort
  • Stretchable polyurethane coating on the palm for grip capabilities
  • Antistatic properties offer the user hand protection against electrostatic discharge
  • Low lint for cleanroom applications
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Construction Seamless Knit
Coating Material Polyurethane
Color Gray
Hand Specific
Material Polyamide
Product Type Anti-static Gloves
Size 9
EN388 Standard 3131A
Gender Unisex
Glove Cuff Style Knit
Palm Type Coated
Series 11-130
Yarn Gauge 13