Learn More
Ansell HyFlex™ 11-135 Seamless Knit Polyamide Antistatic Gloves with Coated Fingertips
Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
Description
- Gloves are knitted using a seamless, non-stop process
- Textile liner made of high-tenacity polyamid continuous thread that includes copper
- Seamless, fine stitches enhance user comfort
- Stretchable polyurethane coating on the fingertips for grip capabilities
- Antistatic properties offer the user hand protection against electrostatic discharge
- Low lint for cleanroom applications
Specifications
Specifications
| Certifications/Compliance (Europe) | EN 420:2003 + A1:2009 |
| Construction | Seamless Knit |
| Coating Material | Polyurethane |
| Color | Gray |
| Exterior Finish | No Texture |
| Hand | Specific |
| Material | Polyamide |
| Product Type | Anti-static Gloves |
| Size | 7 |
| EN388 Standard | 113XA |
| Show More |