Learn More
Ansell HyFlex™ 11-135 Seamless Knit Polyamide Antistatic Gloves with Coated Fingertips
Product Code. 16069591 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
Unit Size:
12 pairs
Product Code. 16069591
Brand: Ansell 11135070
Only null left
Add to basket
Add to basket
Gloves are knitted using a seamless, non-stop process with high-tenacity polyamid continuous thread. Antistatic properties protect users from electrostatic discharge.
  • Gloves are knitted using a seamless, non-stop process
  • Textile liner made of high-tenacity polyamid continuous thread that includes copper
  • Seamless, fine stitches enhance user comfort
  • Stretchable polyurethane coating on the fingertips for grip capabilities
  • Antistatic properties offer the user hand protection against electrostatic discharge
  • Low lint for cleanroom applications
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 420:2003 + A1:2009
Construction Seamless Knit
Coating Material Polyurethane
Color Gray
Exterior Finish No Texture
Hand Specific
Material Polyamide
Product Type Anti-static Gloves
Size 7
EN388 Standard 113XA
Gender Unisex
Glove Cuff Style Knit
Series 11-135
Yarn Gauge 13