Learn More
Ansell HyFlex™ 11-725 Polyurethane-Coated Cut-Resistant Lycra™ Gloves
Product Code. 16679073 Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Product Code. 16679073
Brand: Ansell 11725100
Only null left
Add to basket
Edge
Add to basket
Gloves combine comfort and dexterity with a high degree of cut resistance for applications that involve sharp objects.
  • A balance of comfort, dexterity, and sure handling
  • High level of cut resistance for safe use in applications involving contact with sharp piece
  • Intercept™ is a specialized knitting technology that blends engineered, synthetic, and natural fibers into high performance yarns
  • Lycra™ in the liner/coating provides enhances dexterity and fit
  • Designed for use in dry, as well as slightly oily environments
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Polyurethane
Color White
Cut Resistance Rating ANSI A3
Hand Specific
Latex Free No
Material Intercept Technology Yarn
Product Type Mechanical Protection Gloves
Size 10
EN388 Standard 4342B
For Use With (Application) Automotive Assembly, Construction, Maintenance, and Metal Fabrication
Glove Cuff Style Knit
Series 11-725
Yarn Gauge 15