Learn More
Ansell™ HyFlex 11-840 Gloves
Click to view available options
Size:
5
12
Description
- These industrial hand gloves offer Fortix™ Abrasion Resistance Technology for enhanced durability
- Ideal nitrile-coated work gloves with up to 2x the grip for safer handling
- Enhanced breathability for even better comfort
Specifications
Coating Material | Nitrile Foam |
Color | Black, Gray |
Cut Resistance Rating | ANSI A1 |
Hand | Ambidextrous |
Latex Free | Yes |
Material | Nylon and Spandex |
Size | 12 |
Style | 11-840 |
Glove Cuff Style | Knit |
Length (Metric) | 200 to 275 mm |