Learn More
Thermo Scientific™ HyperSep™ Retain PEP SPE 96-Well Plates and Individual Wells
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP SPE 96-Well Plates and Individual Wells.
Brand:  Thermo Scientific™ 60303-202
Additional Details : Weight : 0.30000kg
Product Code. 10742327
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products »
Description
Description
Get balanced retention for polar and nonpolar analytes with Thermo Scientific™ HyperSep™ Retain PEP SPE 96-Well Plates and Individual Wells, packed with a high capacity, high purity, highly porous polystyrene DVB material modified with urea functional groups. With chemistry pH stable 0 to 14, the of these cartridges in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Features: Ideal for large samples, Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • [bullet] High, consistent recoveries
  • [bullet] Fast and easy sample preparation and method development
  • [bullet] Reproducible, reliable sample preparation in a traditional format
  • [bullet] Available in a wide range of chemistries and bed weights
Applications:
  • [bullet] Drugs and metabolites in biological fluids
  • [bullet] Peptides in serum, plasma or biological fluids
  • [bullet] Environmental samples
For Use With: The Thermo Scientific™ HyperSep-96™ System
Specifications
Specifications
Item Description
30 to 50 μm
Individual Well
Screening of Drugs, Metabolites, Peptides in Biological Samples
HyperSep Retain PEP