Läs mer
Thermo Scientific™ HyperSep™ Tip Microscale SPE Extraction Tips, 10-200μL
Handla allt< titel> ProdukterBeskrivning
Get faster sample preparation with minimal sample loss with these revolutionary SPE micropipette tips and achieve separation in volumes as low as 100nL.
Thermo Scientific™ HyperSep™ Microscale SPE Tips are available in many popular chemistries to suit a wide range of applications. They feature a unique micropipette tip that has the chromatographic material directly attached to its inner surface. No glues or polymers are used, so there's no contamination from the supporting matrix.
Mass spectrometry
Desalting
Protein purification
MALDI
Electrophoresis
HPCE
HPLC
CEC
Tips are available in 1-10µL capacity and 10-200µL capacity.
Specifikationer
Specifikationer
| Description | Titanium Dioxide; 10 to 200 uL Volume |
| For Use With (Application) | Microscale Solid Phase Extraction |
| For Use With (Equipment) | Micropipettes |
| Product Line | HyperSep |
| Sorbent | HyperSep Titanium Dioxide |
| Type | SPE Tips |
| Volume (Metric) | 10 to 200 μL |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.