Learn More
Thermo Scientific™ Hypersil™ Silica Drop-in Guard Cartridges
Product Code. 10038992 Shop All Thermo Scientific Products
Change view
Click to view available options
:
2.1 x 10mm ID x L; 5μm particle size
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10038992 2.1 x 10mm ID x L; 5μm particle size
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10038992 Supplier Thermo Scientific™ Supplier No. 30005012101
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Convenience, economy and effective protection for extending column lifetime.
Thermo Scientific™ Hypersil Silica Drop-in Guard Cartridges offer convenience, economy and effective protection for extending column lifetime.
  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders
  • Available in packs of 4
Requires:
Cartridge holder
TRUSTED_SUSTAINABILITY
Specifications
Product Line Hypersil Classical