Learn More
Axygen™ I-4010 Block Casing
Product Code. 17812617 Shop All Corning Products
Click to view available options
For Use With (Equipment):
Axygen™ I-4010-A, I-4010-B, I-4010-C, I-4010-E
Axygen™ I-4010-H and I-4010-I
Unit Size:
Each
Product Code. 17812617
Brand: Axygen™ I4010TPC
Only null left
Add to basket
Add to basket
Spare block casing for heat preservation while operating the Axygen Microtube Shaker.
  • Spare block casing
TRUSTED_SUSTAINABILITY
Specifications
Weight (Metric) 0.3 kg
For Use With (Equipment) Axygen™ I-4010-A, I-4010-B, I-4010-C, I-4010-E