Learn More
BUCHI I-80 Controller for Rotavapor R-80
Shop All Buchi Products
Click to view available options
For Use With (Equipment):
Rotavapor R-80 House Vacuum
Rotavapor R-80 V-80/180
Includes:
Communication Cable, Holder
Power Pack
Woulff Bottle, Communication Cable, Tubing
Unit Size:
Each
Description
- Functional design fulfills essential needs of vacuum regulation
- Saves resources thanks to control of vacuum pump
- High longevity due to robust and durable design
- User-friendly and intuitive handling due to self-explaining user interface
- Easy and accurate vacuum regulation
- Relevant information is displayed at a glance
- High reliability due to centralized control of major process parameters
- Full process control thanks to parameter monitoring
- Proven technology for major applications
Specifications
Specifications
| Type | Controller Interface |
| For Use With (Equipment) | Rotavapor R-80 House Vacuum |
| Includes | Woulff Bottle, Communication Cable, Tubing |