Learn More
Thermo Scientific™ E. coli Lysis Buffer-II
Product Code. 15434949
Click to view available options
Quantity:
250mL
500mL
Unit Size:
250mL
500mL
Product Code. 15434949
Brand: Thermo Scientific™ J62613.AK
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
Quantity 250mL
Format Liquid
Product Type Cell Lysis Reagent
Content And Storage Ambient temperatures