missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Silencer™ Select Human Long Non-coding RNA (lncRNA) siRNA Library

Product Code. p-9697006
Click to view available options
Format:
384-well Plate
96-well Plate
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15662426

Brand: Invitrogen™ A30148

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The Silencer™ Select Human Long Noncoding RNA (lncRNA) siRNA Library contains 15,081 Ambion™ Silencer Select siRNAs targeting 5,027 lncRNAs as identified in GENCODE v22.

  • Long noncoding RNAs (lncRNAs) have gained interest in recent years as they have been implicated in genomic regulation, cellular housekeeping functions, developmental processes and disease. The Silencer™ Select Human Long Noncoding RNA (lncRNA) siRNA Library contains 15,081 Ambion™ Silencer Select siRNAs targeting 5,027 lncRNAs as identified in GENCODE v22.
  • The Silencer Select Human Long Noncoding RNA (lncRNA) siRNA Library is designed against targeted transcripts aligned to the entire genome, maximizing prediction and coverage across alternatively spliced transcripts. Silencer Select siRNAs are designed with Thermo Fisher Scientific’s leading design algorithms and locked nucleic acid (LNA) chemical modifications to help ensure efficacy, specificity and potency. This in turn provides a powerful platform for achieving accurate and consistent phenotype data from RNAi screens in the discovery and elucidation of lncRNAs.
  • Superior efficacy—Reduce off target effects by up to 90% due to latest design algorithms and LNA chemical modifications
  • Highest efficiency—Perform knockdown studies with 100-fold more potent siRNAs than currently available siRNAs
  • Dependable—Achieve faster discoveries via consistency of gene knockdown and reliability of phenotypic results
  • Convenient—Reduce data de-convolution and accelerate target validation with our 96- and 384-well arrayed format
  •  Customizable—Choose the best option for your workflow demands, including additional siRNA scales and pooled formats
  • Together with careful bioinformatics selection of gene targets, Silencer Select siRNAs are optimized with latest design algorithms, proprietary chemical modifications and high quality synthesis to help ensure desired RNAi outcomes
  • The Silencer Select Human Long Noncoding RNA (lncRNA) siRNA Library contains our pre-designed and when available, validated siRNAs generated with highest potency and specificity in mind so you can focus on the highest data quality backed by the best in industry guarantee
  • Ambion Silencer Select siRNAs are designed with the highest potency of available siRNAs, resulting in 100-fold more potent siRNAs and allowing gene knockdowns at the lowest concentrations (see figure below). The highly potent Silencer Select siRNAs allow you to use even lower concentrations of siRNAs, which helps to minimize off target effects, reduce false negative data and maximize siRNA library output for additional screens and data validation. The Silencer Select Human Long Noncoding RNA (lncRNA) siRNA Library contains 0.25 nmol of each highly potent siRNA, sufficient for at least 500 transfections (at 5 nM siRNA, 100 μL transfection volume)
  • Thermo Fisher Scientific offers a comprehensive list of pre-defined or custom siRNA libraries to human, mouse or rat genes with 1, 2, or 5 nmol of each siRNA or larger sizes.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 15081 total siRNAs targeting 5027 lncRNA transcripts plated onto 96-well plates (174 plates)

Store at or below -20°C in a non-frost-free freezer.
Format 96-well Plate
RNAi Type siRNA
Species Human
Sample Type RNA
For Use With (Application) RNAi
Product Line Silencer™Select
Product Type siRNA Library
Purity Standard
Quantity 1 set (96-well)
Shipping Condition Approved for shipment at Room Temperature or on Wet Ice
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.