missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IBMX (3-isobutyl-1-methylxanthine), MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
Description
- IBMX (3-isobutyl-1-methylxanthine), like other methylated xanthine derivatives, is a competitive non-selective phosphodiesterase inhibitor which raises intracellular cAMP, activates PKA, inhibits TNFα, and leukotriene synthesis
- It reduces inflammation and innate immunity, and nonselective adenosine receptor antagonist
Specifications
Specifications
| Quantity | 1 g |
| Color | White |
| Purity | >99% |
| Molecular Weight (g/mol) | 222.3 |
Safety and Handling
Product Identifier
- 3-Isobutyl-1-methylxanthine
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 3-Isobutyl-1-methylxanthine
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction