missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ICP-MS P/A Tuning Solution 2: Ge, Mo, Pd, Ru, Sb, Sn @ 10 ug/mL; Ir, Ti @ 5 ug/mL in 10% HCl / 1% HNO3 /Tr. HF, VHG™
Description
ICP-MS P/A Tuning Solution 2: Ge, Mo, Pd, Ru, Sb, Sn @ 10 ug/mL; Ir, Ti @ 5 ug/mL in 10% HCl / 1% HNO3 /Tr. HF
Specifications
Specifications
| Chemical Name or Material | Iridium/Titanium/Antimony/Germanium/Molybdenum/Palladium/Ruthenium/Tin |
| CAS | 7439-88-5, 7440-32-6, 7440-36-0, 7440-56-4, 7439-98-7, 5-3-7440 |
| Analyte or Component Names | Iridium/Titanium/Antimony/Germanium/Molybdenum/Palladium/Ruthenium/Tin |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | Dilute HNO3/HF |
| Quantity | 100 mL |
| Concentration or Composition (by Analyte or Components) | Iridium 5 μg/mL/Titanium 5 μg/mL/Antimony 10 μg/mL/Germanium 10 μg/mL/Molybdenum 10 μg/mL/Palladium 10 μg/mL/Ruthenium 10 μg/mL/Tin 10 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?