Learn More
Sampling Systems™ ID Tape
Quickly identify the status of a container to prevent it being used in error. Sampling Systems™ ID Tape allows containers, pallets, and bags to be easily labelled with highly visible tape.
Brand: Sampling Systems™ 8221A-01
Additional Details : Weight : 0.54720kg
Description
- Use with standard tape dispensers
- High strength adhesive
- 10 rolls per box
Specifications
Medium strength adhesion tape | |
Labeled ID Tape | |
10/Pk. | |
66 mm |
Red | |
Quarantine | |
66 x 50 mm | |
50 mm |