missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Ifenprodil hemitartrate

Product Code. 18085842
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18085842

Brand: Tocris Bioscience™ 0545/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Non-competitive NMDA antagonist; also σ ligand

NMDA receptor antagonist, acting at the polyamine site. Also an α-adrenergic vasodilator. σ2 ligand displaying about 3-fold selectivity over σ1 sites.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23210-58-4

Specifications

Quantity 50 mg
Molecular Formula C46H60N2O10
Formula Weight 400.49
Purity >99%
Molecular Weight (g/mol) 800.99
Chemical Name or Material Ifenprodil hemitartrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.