Learn More
IGF2BP1 Antibody, Novus Biologicals™
Code produit 18620656 Tous les produits Bio Techne Produits
Modifier l'affichage
Click to view available options
Quantité:
25 μL
0.1 mL
Conditionnement:
0.10mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantité Conditionnement
18620656 25 μL 25µL
18095186 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 18620656 Fournisseur Novus Biologicals Code fournisseur NBP23895625ul
 Article restreint
Estimated Shipment: 02-12-2026
Ajouter au panier
Ajouter au panier
missing translation for 'validMainframeMsg'
Rabbit Polyclonal Antibody
IGF2BP1 Polyclonal specifically detects IGF2BP1 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Antigène IGF2BP1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugué Unconjugated
Dilution Western Blot 0.04 - 0.4 μg/mL, Simple Western 1:10, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formule PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Numéro d’ordre du gène Q9NZI8
Alias de gène Coding region determinant-binding protein, CRD-BP, CRDBPIGF-II mRNA-binding protein 1, IGF II mRNA binding protein 1, IMP1, IMP-1ZBP-1, insulin-like growth factor 2 mRNA binding protein 1, insulin-like growth factor 2 mRNA-binding protein 1, VICKZ family member 1, VICKZ1, ZBP1IGF2 mRNA-binding protein 1, Zip code-binding protein 1, Zipcode-binding protein 1
Symboles de gène(s) IGF2BP1
Espèces hôtes Rabbit
Immunogène This antibody was developed against a recombinant protein corresponding to amino acids: HALKVSYIPDEQIAQGPENGRRGGFGSRGQPRQGSPVAAGAPAKQQQVDI
Méthode de purification Affinity Purified
Quantité 25 μL
État réglementaire RUO
Primaire ou secondaire Primary
Identification génétique (Entrez) 10642
Spécificité du test Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Espèces cibles Human
Contenu et stockage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG