missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IgG(H+L) Donkey anti-Rabbit, AP, Secondary Antibody, Proteintech
Donkey anti-Rabbit Affinipure Secondary Antibody
50.00€ - 179.00€
Specifications
| Antigen | IgG(H+L) |
|---|---|
| Applications | ELISA, Western Blot |
| Conjugate | Alkaline Phosphatase (AP) |
| Form | Liquid |
| Host Species | Donkey |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17690317
|
Proteintech
SA00002-5-200UL |
200 μL |
50.00€
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17680317
|
Proteintech
SA00002-5-1ML |
1 mL |
179.00€
1mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| IgG(H+L) | |
| Alkaline Phosphatase (AP) | |
| Donkey | |
| 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
| The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. |
| ELISA, Western Blot | |
| Liquid | |
| Rabbit | |
| Secondary | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title