missing translation for 'onlineSavingsMsg'
Learn More

IgG(H+L) Goat anti-Human, AP, Secondary Antibody, Proteintech

Product Code. 17640327 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
17640327 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 17640327

Brand: Proteintech SA0000281ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat anti-Human Affinipure Secondary Antibody

Specifications

Antigen IgG(H+L)
Applications ELISA, Western Blot
Conjugate Alkaline Phosphatase (AP)
Formulation 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Host Species Goat
Purification Method The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Quantity 1 mL
Primary or Secondary Secondary
Target Species Human
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.