Learn More
IgG(H+L) Goat anti-Mouse, Rhodamine, Secondary Antibody, Proteintech
Goat anti-Mouse Affinipure Secondary Antibody
Brand: Proteintech SA00007-1-500UL
This item is not returnable.
View return policy
Specifications
| IgG(H+L) | |
| Rhodamine | |
| Goat | |
| 500 μL | |
| Mouse | |
| Liquid |
| Immunofluorescence, Flow Cytometry | |
| 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA | |
| The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads. | |
| Secondary | |
| Store at -20°C. Stable for one year after shipment. |