Learn More
IgG(H+L) Goat anti-Rabbit, AMCA, Secondary Antibody, Proteintech
Product Code. 17630377 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
200 μL
500 μL
Unit Size:
200µL
500µL
Product Code. Quantity unitSize
17630377 500 μL 500µL
17620377 200 μL 200µL
2 options
Product Code. 17630377
Brand: Proteintech SA000102500UL
Goat anti-Rabbit Affinipure Secondary Antibody
Specifications
Antigen IgG(H+L)
Applications Immunofluorescence, Flow Cytometry
Conjugate AMCA
Formulation 0.01M Sodium phosphate, 0.25M NaCl, 50% glycerol, pH7.6, with 3 mg/mL BSA
Host Species Goat
Purification Method The antibody was purified from antisera by immunoaffinity chromatography using antigens coupled to agarose beads.
Quantity 500 μL
Primary or Secondary Secondary
Target Species Rabbit
Content And Storage Store at -20°C. Stable for one year after shipment.
Form Liquid