Learn More
Rat IgG1 kappa Isotype Control (eBRG1), PE, eBioscience™, Invitrogen™
Rat Isotype Control
108.00€ - 264.00€
Specifications
Antigen | Rat IgG1 kappa |
---|---|
Clone | eBRG1 |
Concentration | 0.2 mg/mL |
Applications | Control, Flow Cytometry |
Conjugate | PE |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15123842
|
Affymetrix eBioscience
12-4301-81 |
50 μg |
108.00€
50µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
13496853
|
Affymetrix eBioscience
12-4301-82 |
100 μg |
180.00€
100µg |
In Stock
Log in to see stock. |
|||||
15228769
|
Affymetrix eBioscience
12-4301-83 |
200 μg |
264.00€
200µg |
In Stock
Log in to see stock. |
|||||
Description
Description: The monoclonal rat IgG1, kappa is useful as an isotype control immunoglobulin. Applications Reported: This rat IgG1 isotype control has been reported for use in flow cytometric analysis. Applications Tested: The PE rat IgG1 isotype control has been tested by flow cytometric analysis of mouse splenocyte suspensions and can be used at the same concntration as the experimental antibody. Excitation: 488-561 nm; Emission: 578 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG1 is a subclass of immunoglobulin G (IgG) antibodies that are commonly used in research and diagnostics. These antibodies are produced by rats in response to an antigen and can be purified from serum or hybridoma cell culture supernatants. Rat IgG1 is often used as a negative control in experiments, as it does not bind to many mammalian antigens. It is also used in immunohistochemistry, flow cytometry, and western blotting to detect specific antigens. Gene targeting is a powerful tool used to manipulate the rat genome and generate animal models for studying human diseases. With gene targeting, specific genes can be knocked out, inserted, or modified to investigate their role in disease pathogenesis.Specifications
Rat IgG1 kappa | |
0.2 mg/mL | |
PE | |
Rat | |
Not Applicable | |
Immunoglobulin G; Immunoglobulin G1; ImmunoglobulinG; ImmunoglobulinG1 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBRG1 | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG1 κ | |
Affinity chromatography |
For Research Use Only.