missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Mouse IgG1 Isotype Control, FITC, Invitrogen™

Mouse Isotype Control

Brand:  Invitrogen™ MG101

92.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.



Additional Details : Weight : 0.50000kg

Product Code. 10373353

  • 138.00€ / Each
In Stock
to see stock.
Add to basket

Description

Description

Mouse IgG1 Isotype Control for Flow

Mouse IgG1 Control antibody specifically detects Mouse IgG1 in Not Applicable samples. It is validated for Flow Cytometry.
Specifications

Specifications

Mouse IgG1
FITC
IgG1; Igh-4; Ighg1; Immunoglobulin G; Immunoglobulin G1; immunoglobulin heavy constant gamma 1 (G1m marker); ImmunoglobulinG; ImmunoglobulinG1; VH7183
purified
ASR
Isotype Control (Primary)
2-8°C
IgG1
Wet Ice
Flow Cytometry
PBS with 0.1% sodium azide
Mouse
500 μL
Whole Antibody
Not Applicable
Liquid
50 Tests
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions