Learn More
Rat IgG2a kappa Isotype Control (eBR2a), APC-eFluor™ 780, eBioscience™, Invitrogen™
Rat Isotype Control
104.00€ - 198.00€
Specifications
Antigen | Rat IgG2a kappa |
---|---|
Clone | eBR2a |
Concentration | 0.2 mg/mL |
Applications | Control, Flow Cytometry |
Conjugate | APC-eFluor 780 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15351650
|
Affymetrix eBioscience
47-4321-80 |
25 μg |
104.00€
25µg |
Estimated Shipment: 06-05-2024
Log in to see stock. |
|||||
15361650
|
Affymetrix eBioscience
47-4321-82 |
100 μg |
198.00€
100µg |
In Stock
Log in to see stock. |
|||||
Description
Description: The monoclonal rat IgG2a, kappa is useful as an isotype control immunoglobulin. Applications Reported: APC-eFluor™ 780 Rat IgG2a Isotype Control (APC-Alexa Fluor™ 750 replacement) has been reported for use in flow cytometric analysis. Applications Tested: Rat IgG2a K Isotype Control APC-eFluor™ 780 has been tested by flow cytometric analysis of mouse splenocytes. Use isotype control at the same concentration as experimental antibody. APC-eFluor 780 emits at 780 nm and is excited with the Red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Light sensitivity: This tandem is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 633-647 nm; Emission: 780 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2a kappa Control antibody specifically detects Rat IgG2a kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.Specifications
Rat IgG2a kappa | |
0.2 mg/mL | |
APC-eFluor 780 | |
Rat | |
Not Applicable | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBR2a | |
Control, Flow Cytometry | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG2a κ | |
Affinity chromatography |
For Research Use Only.