Learn More
Rat IgG2a kappa Isotype Control (eBR2a), eFluor™ 615, eBioscience™, Invitrogen™
Rat Isotype Control
273.00€ - 464.00€
Specifications
Antigen | Rat IgG2a kappa |
---|---|
Clone | eBR2a |
Concentration | 0.2 mg/mL |
Applications | Control, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry |
Conjugate | eFluor 615 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15371100
|
Affymetrix eBioscience
42-4321-80 |
25 μg |
273.00€
25µg |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
15381100
|
Affymetrix eBioscience
42-4321-82 |
100 μg |
464.00€
100µg |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
Description: The monoclonal rat IgG2a is useful as an isotype control immunoglobulin. Applications Reported: Rat IgG2a Isotype Control eFluor® 615 has been reported for use in immunohistochemical and immunocytochemical staining. Applications Tested: Rat IgG2a Isotype Control eFluor® 615 has been tested by immunocytochemistry or immunohistochemistry. Use at the same concentration as the experimental antibody. This product has not been validated for flow cytometric analysis. Filter Recommendation: When using this eFluor® 615 antibody conjugate, we recommend a filter that will capture the 615 emission wavelength (for example, Excitation 560/55, 585LP, Emission 645/75). A standard Alexa Fluor® 594 filter is acceptable. Excitation: 595 nm; Emission: 615 nm. Filtration: 0.2 μm post-manufacturing filtered.
Rat IgG2a kappa Control antibody specifically detects Rat IgG2a kappa in Not Applicable samples. It is validated for Control, Immunocytochemistry, Immunohistochemistry.Specifications
Rat IgG2a kappa | |
0.2 mg/mL | |
eFluor 615 | |
Rat | |
Not Applicable | |
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2 | |
Isotype Control (Primary) | |
4° C, store in dark, DO NOT FREEZE! |
eBR2a | |
Control, Immunocytochemistry, Immunohistochemistry, Immunohistochemistry | |
Liquid | |
RUO | |
PBS with 0.09% sodium azide; pH 7.2 | |
IgG2a κ | |
Affinity chromatography |
For Research Use Only.