missing translation for 'onlineSavingsMsg'
Learn More

Mouse IgG2b kappa Isotype Control (eBMG2b), eFluor™ 450, eBioscience™, Invitrogen™

Mouse Isotype Control

Brand:  Affymetrix eBioscience 48-4732-80

78.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.



Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15381770

  • 117.00€ / 25µg
In Stock
to see stock.
Add to basket

Description

Description

Description: The monoclonal mouse IgG2b, kappa is useful as an isotype control immunoglobulin. Applications Reported: Mouse IgG2b Isotype Control eFluor® 450 has been reported for use in flow cytometric analysis. Applications Tested: eFluor® 450 Mouse IgG2b Isotype Control has been tested by flow cytometric analysis of normal human peripheral blood cells. Use isotype control at the same concentration as experimental antibody. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 μm post-manufacturing filtered.

Mouse IgG2b kappa Control antibody specifically detects Mouse IgG2b kappa in Not Applicable samples. It is validated for Control, Flow Cytometry.
Specifications

Specifications

Mouse IgG2b kappa
eBMG2b
eFluor 450
IgG2; Immunoglobulin G; Immunoglobulin G2; ImmunoglobulinG; ImmunoglobulinG2
Affinity chromatography
RUO
Not Applicable
Liquid
Control, Flow Cytometry
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
Mouse
25 μg
Isotype Control (Primary)
4° C, store in dark, DO NOT FREEZE!
IgG2b κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only.