Learn More
Invitrogen™ Mouse IgG2b Uncoated ELISA Kit
Description
The Mouse Immunoglobulin G2b (IgG2b) Uncoated ELISA Kit contains pre-matched antibody pairs, plates, and reagents for performing quantitative enzyme linked immunosorbent assays (ELISA) to detect and quantify protein levels of mouse IgG2b. Wash Buffer and Stop Solution are needed to complete the ELISA reaction and are sold separately. Principle of the method ELISAs are designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody is coated to the bottom of the wells of a microplate, which is an overnight process. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen.
Specifications
Specifications
| Assay Range | 3.90-250 ng/mL |
| Assay Sensitivity | 3.90 ng/mL |
| Content And Storage | Capture Antibody ConcentrateDetection Antibody ConcentrateMouse IgG2b isotype control (standard), lyophilized, 500 ng/mL upon reconstitutionPhosphate Buffered Saline Concentrate (PBS)Assay Buffer A Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)Tetramethylbenzidine (TMB) Substrate Solution96-well plates included with product Cat. Nos. ending in suffixes -22, -76, -86 |
| Conjugate | HRP |
| Immunoassay Kit Format | Uncoated ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Mouse |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.