missing translation for 'onlineSavingsMsg'
Learn More

IgM Rat anti-Mouse, Biotin, Clone: eB121-15F9, eBioscience™

Rat Monoclonal Antibody

Marque:  Affymetrix eBioscience 13-5890-81

Informations supplémentaires : Poids : 0.25000kg

 Afficher plus de versions de ce produit

Code produit. 15279899

  • 62.75€ / 50µg


Date dexpédition estimée
du fournisseur 24-10-2019
Ajouter au panier

Description

Description

IgM (Immunoglobulin M) is expressed intracellularly during the early stages of B lymphopoiesis, and then on the surface of more mature B cells in the bone marrow and peripheral B cells. The isotype of a primary antibody and its application can result in background staining. Primary antibody background noise can be caused by binding to Fc receptors on target cells; by non-specific interactions with cellular proteins, carbohydrates, and lipids; or by cell autofluorescence. Isotype control antibodies can act as negative controls to help differentiate non-specific background signal from specific antibody signal because they have no relevant specificity to a target antigen. While isotype controls are most commonly used in flow cytometry, they are also useful in other applications such as chromatin immunoprecipitation (ChIP), immunohistochemistry, and gel shifts. Isotype controls should match with the primary antibody species and isotype so that the level of specific staining by the primary antibody may be accurately determined. If using directly labeled primary antibodies, the isotype control works best if conjugated with the same label as the test antibody.
Spécification

Spécification

IgM
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
Liquid
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
eB121-15F9
Biotin
Ighm
Ighm
IgG2a, kappa
50 μg
4° C, do not freeze
Primary
16019
Vidéos
Documents

For Research Use Only.