Learn More
Proteintech IHCeasy MAdCAM1 Ready-To-Use IHC Kit
A ready-to-use IHC kit for the staining of MAdCAM1.
Brand:  Proteintech KHC0025-50T
Additional Details : Weight : 0.55700kg Transport : UN number : 3316 Chem class : 9 Pack group : III
Product Code. 17436733
  • 389.00€ / 1 set
Estimated Shipment: 17-05-2024
to see stock.
Description
Description
  • This kit provides all reagents, from antigen retrieval to cover slip mounting, that require little to no diluting or handling prior to use.
  • Simply apply the reagents to your sample slide according to the protocol and you're steps away from obtaining high-quality IHC data.
Specifications
Specifications
Human
Store at 2°C to 8°C
50 Reactions
FFPE Tissue
50 Tests