missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ IL-16 Antibody Pair [HRP]

Product Code. 18610287 Shop All Bio Techne Products
Click to view available options
Quantity:
15 Plates
5 Plates
Unit Size:
Pack of 15
Pack of 5
This item is not returnable. View return policy

Product Code. 18610287

Brand: Novus Biologicals™ NBP31165615Plates

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Pair

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 3.13 to 200 pg/mL (example only, lot dependent)
Assay Sensitivity 3.13 pg/mL (example only; lot dependent)
Conjugate HRP
Detection Method Sandwich ELISA
Immunoassay Kit Format Sandwich ELISA Kit
Product Type Sandwich ELISA
Specificity This solid phase sandwich ELISA utilizes a monoclonal antibody specific for Mouse IL-16.
Target Species Mouse
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 3603
Gene Symbol IL16
Quantity 15 Plates
Research Discipline Biologically Active Proteins, Cancer, Cytokine Research, Hypoxia, Immunology, Innate Immunity
Storage Requirements Storage of components varies. See protocol for specific instructions.
Synonym FLJ16806, FLJ42735, FLJ44234, HsT19289, IL-16, interleukin 16 (lymphocyte chemoattractant factor), LCFprIL-16, lymphocyte chemoattractant factor, neuronal interleukin 16, NIL16, PRIL16, prointerleukin 16, pro-interleukin-16
Target Mouse
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.