Learn More
Invitrogen™ IL-36G (IL-1F9) Human ELISA Kit
Sandwich ELISA Kit
668.00€ - 2635.00€
Specifications
Accession Number | Q9NZH8 |
---|---|
Assay Sensitivity | 13 pg/mL |
Assay | Solid-phase sandwich ELISA |
Conjugate | Biotin |
Product Type | ELISA Kit |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15465043
|
Invitrogen™
EHIL36G |
96 Tests |
668.00€
1 set |
Estimated Shipment: 06-06-2024
Log in to see stock. |
|||||
17145489
|
Invitrogen™
EHIL36GX5 |
5 x 96 Tests |
2635.00€
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
The Human Interleukin 36G (IL-36G/IL-1F9) ELISA quantitates Hu IL-36G in human serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-36G. Principle of the method The Human IL-36G solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Cytokine that binds to and signals through the IL1RL2/IL-36R receptor which in turn activates NF-kappa-B and MAPK signaling pathways in target cells. Part of the IL-36 signaling system that is thought to be present in epithelial barriers and to take part in local inflammatory response; similar to the IL-1 system with which it shares the coreceptor IL1RAP. Seems to be involved in skin inflammatory response by acting on keratinocytes, dendritic cells and indirectly on T-cells to drive tissue infiltration, cell maturation and cell proliferation. In cultured keratinocytes induces the expression of macrophage, T-cell, and neutrophil chemokines, such as CCL3, CCL4, CCL5, CCL2, CCL17, CCL22, CL20, CCL5, CCL2, CCL17, CCL22, CXCL8, CCL20 and CXCL1; also stimulates its own expression and that of the prototypic cutaneous proinflammatory parameters TNF-alpha, S100A7/psoriasin and inducible NOS. May play a role in proinflammatory responses during particular neutrophilic airway inflammation: activates mitogen-activated protein kinases and NF-kappa B in primary lung fibroblasts, and stimulates the expression of IL-8 and CXCL3 and Th17 chemokine CCL20 in lung fibroblasts. May be involved in the innate immune response to fungal pathogens, such as Aspergillus fumigatus.Specifications
Q9NZH8 | |
Solid-phase sandwich ELISA | |
ELISA Kit | |
Plasma, Serum, Supernatant | |
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis | |
56300 | |
<10% | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |
13 pg/mL | |
Biotin | |
Other Proteins | |
Human | |
Colorimetric Microplate Reader | |
<12% | |
HRP | |
Inflammation | |
Human | |
4 hr. 45 min. |