missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-6R/IL-6RA Mouse ELISA Kit

Sandwich ELISA Kit

Brand:  Invitrogen™ EMIL6RAX5

Additional Details : Weight : 0.33800kg

 View more versions of this product

Product Code. 17185749

  • 2635.00€ / Each
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

The Mouse Interleukin-6 RA (Ms IL-6R/IL-6RA) ELISA quantitates Ms IL-6RA in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms IL-6RA. Principle of the method The Mouse IL-6RA solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

IL6R (interleukin-6 receptor subunit alpha) is a part of the receptor for interleukin 6. The IL-6 receptor complex is composed of two membrane glycoproteins: the low affinity receptor and the signaltransducing component. Dysregulated production of IL6 and this receptor are implicated in the pathogenesis of many diseases, such as multiple myeloma, autoimmune diseases and prostate cancer.
Specifications

Specifications

P22272
6 pg/mL
Solid-phase sandwich ELISA
ELISA Kit
Plasma, Serum, Supernatant
5 x 96 assays
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
16194
<12%
Standard, Wash Buffer, Chromogen, Biotinylated Detection Antibody, Assay Diluent Concentrate, Pre-coated 96 well plates, SAV-HRP, Adhesive Plate Covers, Stop Solution
5 x 96 Tests
Immunology
2°C to 8°C
Mouse
4 hr. 45 min.
6.14 pg/mL to 1500 pg/mL
6 pg/mL to 1500 pg/mL
Biotin
Cytokines & Receptors
This ELISA kit shows no cross-reactivity with the following cytokines tested: Mouse CD30, L CD30, T CD40, CRG-2, CTACK, CXCL16, Eotaxin , Eotaxin-2, Fas Ligand, Fractalkine, GCSF, GM-CSF, IGFBP-3, IGFBP-5, IGFBP-6, IL-1 alpha, IL-1 beta, IL-2, IL-3 Rb, IL-4, IL-5, IL-9, IL-10, IL-12 p70, IL-13, IL-17, KC, Leptin R, LEPTIN(OB), LIX, L-Selectin, Lymphotactin, MCP-1, MCP-5, M-CSF, MIG, MIP-1 alpha, MIP-1 gamma, MIP-2, MIP-3 beta, MIP-3 alpha, PF-4, P-Selectin, RANTES, SCF, SDF-1 alpha, TARC, TCA-3, TECK, TIMP-1, TNF-alpha, TNF RI, TNF RII, TPO, VCAM-1, VEGF
Mouse
Colorimetric Microplate Reader
CD126,IL-6 R,IL-6 R-alpha,IL-6 RA,Il6r
<10%
HRP
RUO
Plasma, 5 μ;L; Serum, 5 μ;L; Supernatant, 100 μ;L
CD126, IL-6 R, Il6r
1 hr. 20 min.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions