missing translation for 'onlineSavingsMsg'
Learn More

IL8 Recombinant Human Protein, hIgG1-Fc Tag

IL8 Recombinant Human Protein, hIgG1-Fc Tag

Marque:  Invitrogen 10098H01H2U25

Informations supplémentaires : Poids : 0.01000kg

 Afficher plus de versions de ce produit

Code produit. 15274635

  • 164.00€ / 25µg


nous consulter 056 260 260
Ajouter au panier

Description

Description

Interleukin 8 (IL8), recombinant human protein is supplied as a lyophilized powder.

This recombinant protein was expressed from a DNA sequence encoding the 72 amino acid residue form (Ser 28-Ser 99) of mature human IL8 (NP_000575.1) fused to the Fc region of human IgG1 at the N-terminus.

Activity: This recombinant protein has not been tested.

Formulation: 140 mM NaCl, 2.7 mM KCl, 10 mM Na2HPO4, 1.8 mM KH2PO4, pH7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween80.

Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2 mg/ml or lower. It is recommended that the protein be aliquoted and be used as soon as possible. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.

Expiration Date: Expires one year from date of receipt when stored as instructed.



This protein is manufactured by SINO Biological.

Spécification

Spécification

Protein assays and analysis
Lyophilized
35kDa
PAGE
5 x 5μg
For research use only. Not for use in diagnostic procedures
CXCL8, GCP-1, GCP1, LECT, LUCT, LYNAP, MDNCF, MONAP, NAF, NAP-1, NAP1
5 Vials of lyophilized protein
Assay standard
Mammalian
Human
Lyophilized
3576
IL8 Recombinant Human Protein
≥85% by SDS-PAGE
–20°C
5 Pack(s)
IL8 Recombinant Human Protein
IL8
Chemokines and receptors
IL8 Recombinant Human Protein
IgG1 Fc Tag, hIgG1-Fc Tag
Vidéos
Documents

For Research Use Only. Not for use in diagnostic procedures.