Learn More
Cytiva ExoProStar DNA and Sequence Reaction Clean-up
Shop All Cytiva Products
Click to view available options
No. of Reactions:
20
100
500
2000
5000
Unit Size:
Each
Beschreibung
Enzymes optimized to work together for high efficiency removal of unincorporated primers and dNTPs. Enzymes provided in two separate tubes, just two simple pipetting steps are needed to prepare the reaction.
- Fast 30-minute protocol
- Scalable for different reaction sizes
- No loss of PCR product
- Easy to automate
- Complete heat inactivation of the enzymes within 15 minutes
Spezifikation
Spezifikation
| Description | Enzymatic PCR clean up technology with illustra Exonuclease I and Alkaline Phosphatase to remove unincorporated primers and dNTPs. For maximum flexibility comes in two separate tubes. |
| For Use With (Application) | Optimized to purify DNA and sequencing set up reactions quickly, efficiently and reliably |
| Includes | One tube of Alkaline Phosphatase and Exonuclease I |
| No. of Reactions | 100 |
| Product Type | illustra ExoProStar |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur