Learn More
Cytiva plasmidPrep Midi Flow Kit
For the rapid extraction and purification of high-quality plasmid DNA from medium scale cultures for use in transfection, sequencing, and enzymatic amplification and modification
Brand:  Cytiva 28-9042-67
Additional Details : CAS : 9048-46-8 Weight : 0.50000kg Transport : UN number : 2790 Chem class : 8 Pack group : III
Product Code. 10166174
  • 804.00€ / Each
Estimated Shipment: 24-07-2024
to see stock.
Includes: Fast-flow plasmid 250 columns; Resuspension, lysis, neutralization, wash, elution, and TE buffers; Full protocol booklet with a detachable, quick reference protocol card
Description
Description
The kit consists of a high-performance, anion-exchange purification medium that combines excellent flow characteristics with an exceptional capacity and specificity for plasmid DNA.
  • High column capacity allows for a proportional yield for input sample volume of 25 to 50mL (for high copy number plasmid) and up to 500mL for low copy number plasmid
  • Provides plasmid DNA with excellent purity and low endotoxin levels
  • Increased ease of use due to minimal pipetting volume changes, color-coded kit components, and pre-equilibrated columns
Specifications
Specifications
illustra plasmidPrep Midi Flow Kit is designed for the purification of high yields of transfection-grade plasmid DNA. The kit allows for the purification of large yields of plasmid DNA that are proportional to the input sample volume.
Fast-flow genomic 250 columns, lyophilized Proteinase K powder, Lysis solutions, load and Elution Buffers, NAP-25 desalting column, spin column adapters, full protocol booklet, detachable, quick reference protocol card
illustra plasmidPrep Midi Flow Kit
150 mL Culture
Used For the rapid extraction and purification of high-quality plasmid DNA from medium scale cultures for use in transfection, sequencing, and enzymatic amplification and modification
25 Preps
1.8
Approx.215 min
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3