Learn More
Cytiva Tissue and Cells genomicPrep Mini Spin Kit
Product Code. 10685545 Shop All Cytiva Products
Change view
Click to view available options
No. of Reactions:
250 Preps
50 Preps
Unit Size:
250 reactions
50 reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. No. of Reactions unitSize
10685545 50 Preps 50 reactions
10714547 250 Preps 250 reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10685545 Supplier Cytiva Supplier No. 28904275
Includes
Spin columns prepacked with a novel silica-membrane; Lyophilized proteinase K powder; Lysis solutions, wash and elution buffers; 2mL Microcentrifuge collection tubes; Full protocol booklet with a detachable, quick reference protocol card
Designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from a variety of animal tissues and mammalian cell cultures
To minimize the incidence of DNA shearing during extraction, the illustra tissue and cells genomicPrep Mini Spin Kit includes a lysis solution with proteinase K to gently release genomic DNA from tissues or cells. The kit can be used to extract DNA from diverse tissue types such as liver, kidney, and mouse tails with yields ranging from 0.5 to 1.5μg of genomic DNA per mg of tissue. The kit consistently generates purified genomic DNA with optical density A260/280 readings of about 1.8.
  • Capable of purifying up to 1.5μg/mg of tissue from an input tissue sample range of 5 to 50mg, and up to 40μg of genomic DNA per 5 million cells depending on cell type
  • Total DNA extraction time reduced significantly to 90 minutes (including tissue disruption)
  • Produces high-quality genomic DNA with high molecular weight and less degradation
  • Validated in several downstream applications including real-time PCR, endpoint PCR, multiplex PCR, and restriction enzyme digests
TRUSTED_SUSTAINABILITY
Specifications
Description illustra tissue and cells genomicPrep Mini Spin Kit is designed for the rapid extraction and purification of high molecular weight genomic DNA (gDNA) from a variety of animal tissues and mammalian cell cultures in just 90 min for tissues and 45 min for ce
For Use With (Application) Used for several downstream applications including—restriction enzyme analysis, ligation, cloning, DNA sequencing and PCR
Includes Spin columns prepacked with a novel Silica membrane, Lyophilized Proteinase K powder, Lysis solution, wash and Elution Buffers, microcentrifuge collection tubes, a full protocol booklet and a detachable, quick reference protocol card
No. of Reactions 50 Preps
Product Type illustra tissue and cells genomicPrep Mini Spin Kit
Purity 1.75
Sample Volume 5-50 mg
Test Time 90 min[Animal Tissue], 45 min[Cultured cells]
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1