Learn More
Motic™ Images Multi Focus Professional Edition (for PC only)
Get the most from your Motic™ microscope system with purposeful and thoughtfully designed software from Motic™. Motic™ Images Multi Focus Professional Edition (for PC only) captures and combines up to 100 images taken at different focusing levels to produce a single, fully focused image with great depth of field.
Brand: Motic™ SE990794
Additional Details : Weight : 0.01000kg
Description
- Combines up to 100 images taken at different focal depths.
- The complex software algorithm will take into account image shift to create an in-focus image with great depth of field.
- Available as a separate purchase or in the Motic™ Images Advanced package.
- Windows™ XP or 2000 are recommended to run this software.
- Compatible with Motic™ Images Plus 2.0 and integrated into Motic™ Advanced 3.2.
Specifications
Software | |
Image stitching software | |
Windows™ XP or 2000 | |
Microscope/camera/computer |