missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Imidazole, Serva Electrophoresis™
Specifications
Specifications
| CAS | 288-32-4 |
| Melting Point | 89°C to 91°C |
| Density | 500 - 600kg/m3 |
| pH | 9.5 to 11 |
| Ignition Point | 480°C |
| Molecular Formula | C3H4N2 |
| MDL Number | MFCD00005183 |
| Quantity | 100 g |
| Synonym | imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole |
| InChI Key | RAXXELZNTBOGNW-UHFFFAOYSA-N |
| Show More |
Safety and Handling
Product Identifier
- Imidazole
- Danger
- Acute toxicity Category 3
- Skin corrosion/irritation Category 1 A
- Reproductive toxicity Category 2
- H301-Toxic if swallowed.
- H314-Causes severe skin burns and eye damage.
- H361d-Suspected of damaging the unborn child.
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?