missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, Pure, Fisher Chemical™

Catalog Number p-7073089
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
288-32-4
C3H4N2
68.08
MFCD00005183
RAXXELZNTBOGNW-UHFFFAOYSA-N
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
795
CHEBI:16069
1H-imidazole
N1C=CN=C1
This item is not returnable. View return policy
288-32-4
C3H4N2
68.08
MFCD00005183
RAXXELZNTBOGNW-UHFFFAOYSA-N
imidazole, glyoxaline, imidazol, iminazole, miazole, 1,3-diazole, glyoxalin, imutex, 1,3-diaza-2,4-cyclopentadiene, pyrro b monazole
795
CHEBI:16069
1H-imidazole
N1C=CN=C1
Melting Point 86°C
Color White to Yellow
pH 10.5
Boiling Point 255°C
Packaging Plastic powder jar
Quantity 100 g
Formula Weight 68.08g/mol
Vapor Pressure 0.003mbar at 20°C
Physical Form Solid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'