missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Immepip dihydrobromide
GREENER_CHOICE

Product Code. 18745477
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18745477

Brand: Tocris Bioscience™ 0932/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Standard H3 agonist. Also H4 agonist

Immepip dihydrobromide is a potent histamine H3 receptor agonist. Also binds to H4 receptors (Ki values are 0.4 and 9 nM at human recombinant H3 and H4 receptors respectively). Equipotent to or slightly more active than (R)-α-methylhistamine at H3 receptors.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(1H-Imidazol-4-ylmethyl)piperidine dihydrobromide
CAS 164391-47-3
Quantity 10 mg
Solubility Soluble to 100mM in water
Molecular Formula C9H15N3.2HBr
Formula Weight Observed MW: 327.06

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.