Learn More
Memmert Incubator IN75mplus TwinDISPLAY, 74L, 20-80°C, Natural Convection, Medical Device
Brand:  Memmert IN75MPLUS
Incubator IN75mplus TwinDISPLAY, 74L, 20-80°C, Natural Convection, Medical Device
Regulatory: This product is under eco-contribution tax.
Product Code. 15634119
  • ONLINE EXCLUSIVE PRICE
    3180.00€
    3079.00€ / Each
    Save 101.00€
    3% Off
Only null left
Estimated Shipment: 05-03-2026