Learn More
3M™ Freeze Watch™ Indicator 9805FW
Product Code. 17288510
Click to view available options
Includes:
400 /Cs.
Unit Size:
Pack of 400
Product Code. 17288510
Brand: 3M™ Freeze Watch™ 7000002736
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance.
View alternative products
Consists of a highly sensitive indicating liquid inside a specially designed ampoule to monitor exposure of temperature-sensitive products to freezing temperatures.
  • Inexpensive solution for monitoring product exposure to freezing temperatures.
  • Self-adhesive backing sticks securely to most exterior packaging materials.
  • Easily-interpreted visual results.
  • Activation temperature of 0°C/32°F helps proper shipping of freeze-sensitive items.
Specifications
Includes 400 /Cs.
Length (English) 2.283 in.
Length (Metric) 58 mm
Temperature (English) Operating 32°F
Temperature (Metric) Operating 0°C